Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech's WDR32 antibody 20483-1-AP primarily detects WDR32 isoform 1 with a theoretical MW of 61 kDa.
Immunogen sequence: TRFLMRMRL TPDCSKMLIS TSSGYLLILH DLDLTNSLEV GSYPILRARR TTSSSGVSPR NSLEVVTPEV LGESDHGNCI TSLQLHPKGW ATLLRCSSNS DDEECTCVYE FQEGAPVRPV SPRCSLRLTH YIEEANVGRG YIKELCFSPD GRMISSPHGY GIRLLGFDKQ CSELVDCLPK EASPLRVIRS LYSHNDVVLT TKFSPTHCQI ASGCLSGRVS LYQPKF (1-225 aa encoded by B C003520)
DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDB1- and CUL4-associated factor 10; WD repeat domain 32; WD repeat-containing protein 32
Gene Aliases: AA959934; DCAF10; RGD1304730; WDR32
UniProt ID: (Human) Q5QP82, (Mouse) A2AKB9
Entrez Gene ID: (Human) 79269, (Mouse) 242418, (Rat) 313242
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support