Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RASGDYDND CTNPITPLCT QPDQVIKGGA SIIQCHILND KRHILTKDTN NNVAYWDVLK ACKVEDLGKV DFEDEIKKRF KMVYVPNWFS VDLKTGMLTI TLDESDCFAA WVSAKDAGFS SPDGSDPKLN LGGLLLQALL EYWPRTHVNP MDEEENEVNH VNGEQENRVQ KGNGYFQVPP HTPVIFGEAG GRTLFRLLCR DSGGETESML LNETVPQWVI DITVDKNMPK FNKIPFYLQP HASSGAKTLK KDRLSASDML QVRKVMEHVY EKIINLDNES QTTSSSNNEK PGEQEKEEDI AVLAEEKIEL LCQDQVLDPN MDLRTVKHFI WKSGGDLTLH YRQKST (158-502 aa encoded by BC037168)
WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids, which commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms involving signal transduction, apoptosis, transcriptional regulation and cell cycle control. WD repeats serve as sites for protein-protein interaction and some seem to mediate the assembly of protein complexes. With eight WD repeats, WDR48 (WD repeat-containing protein 48), also known as USP1-associated factor 1 and p80, is a 677 amino acid protein that functions to regulate deubiquitinating complexes via activation of USP1, USP12 and USP46. WDR48 enhances deubiquitination by increasing catalytic turnover without increasing the affinity of deubiquitinating enzymes for the substrate. WDR48 is ubiquitously expressed and is mainly localized to the cytoplasm. There are five isoforms of WDR48 that are expressed as a result of alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p80; testicular tissue protein Li 224; USP1 associated factor 1; USP1-associated factor 1; WD repeat endosomal protein; WD repeat-containing protein 48
Gene Aliases: 8430408H12Rik; KIAA1449; mKIAA1449; P80; RGD1309702; SPG60; UAF1; WDR48
UniProt ID: (Human) Q8TAF3, (Mouse) Q8BH57
Entrez Gene ID: (Human) 57599, (Rat) 363164, (Mouse) 67561
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support