Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDMRQHEEY IADMALDPAK KLLLTASGDG CLGIFNIKRR RFELLSEPQS GDLTSVTLMK WGKKVACGSS EGTIYLFNWN GFGATSDRFA LRAESIDCMV PVTESLLCTG STDGVIRAVN ILPNRVVGSV GQHTGEPVEE LALSHCGRFL ASSGHDQRLK FWDMAQLRAV VVDDYRRRKK KGGPLRALSS KTWSTDDFFA GLREEGEDSM AQEEKEETGD DSD (162-383 aa encoded by BC002482)
WD repeat domain 55 (WDR55) also named as, FLJ20195, is a 383 amino acid protein, which contains seven WD repeats and belongs to the WD repeat WDR55 family. WDR55 localizes in the nucleus and cytoplasm. WDR55 acts as a modulator of rRNA synthesis and play a central role in organogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat-containing protein 55
Gene Aliases: WDR55
UniProt ID: (Human) Q9H6Y2
Entrez Gene ID: (Human) 54853
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support