Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MHLSSHRLD EYWDRQRNRH RMVKVDPETR YMSLAVCELD QPGLGPLVAA ACSDGAVRLF LLQDSGRILQ LLAETFHHKR CVLKVHSFTH EAPNQRRRLL LCSAATDGSL AFWDLTTMLD HDSTVLEPPV DPGLPYRLGT PSLTLQAHSC GINSLHTLPT REGHHLVASG SEDGSLHVFV LAVEMLQLEE AVGEAGLVPQ LRVLEEYSVP CAHAAHVTGL KILSPSIMVS ASIDQRLTFW RLGHGEPTFM NSTVFHVPDV ADMDCWPVSP EFGHRCALGG QGLEVYNWYD (833-1121 aa encoded by BC002826)
WDR6 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mWDR6; tRNA (34-2'-O)-methyltransferase regulator WDR6; WD repeat domain 6 protein; WD repeat-containing protein 6
Gene Aliases: mWDR6; WDR6
UniProt ID: (Human) Q9NNW5, (Rat) Q5XFW6, (Mouse) Q99ME2
Entrez Gene ID: (Human) 11180, (Rat) 301007, (Mouse) 83669
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support