Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKPESLENSI LDSLEPQSLA SLLSESESPQ EAGRGHPSFL PQQKESSEAS ELILYSLEAE VTVTGTDSQY CRKEVEAGPG DQQGDSYLRV SSDSPKDQSP PEDSGESEAD LECSFAAIHS PAPPPDPAPR FATSLPHFPG CAGPTEDELS LPEGPSVPSS SLPQTPEQEK FLRHHFETLT ESPCRALGDV EASEAEDHFF NPRLSISTQF LSSLQKASRF THTFPPRATQ CLVKSPEVKL MDRGGSQPRA GTGYASPDRT HVLAAGKAEE TLEAWRPPPP CLTSLASCVP ASSVLPTDRN LPTPTSAPTP GLAQGVHAPS TCSYMEATAS SRARISRSIS LGDSEGPIVA TLAQPLRRPS SVGELASLGQ ELQAITTATT PSLDSEGQEP ALRSWGNHEA RANLRLTLSS ACDGLLLPPV DTQPGVTVPA VSFPAPSPVE ESALRLHGSA FRPSLPAPES PGLPAHPSNP QLPEARPGIP GSTASLLEPT SGALGLFQGS PARWSEPWVP VEALPPSPLE LSRVGNILHR LQTTFQEALD LYRVLVSSGQ VDTGQQQART ELVSTFLWIH SQLEAECLVG TSVAPAQALP SPGPPSPPTL YPLASPDLQA LLEHYSELLV QAVRRKARGH
May regulate endogenous transporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: microcephaly, primary autosomal recessive 2; truncated WDR62; WD repeat-containing protein 62
Gene Aliases: C19orf14; MCPH2; WDR62
UniProt ID: (Human) O43379
Entrez Gene ID: (Human) 284403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support