Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EYKILLYSL DGRLLSTYSA YEWSLGIKSV AWSPSSQFLA VGSYDGKVRI LNHVTWKMIT EFGHPAAIND PKIVVYKEAE KSPQLGLGCL SFPPPRAGAG PLPSSESKYE IASVPVSLQT LKPVTDRANP KIGIGMLAFS PDSYFLATRN DNIPNAVWVW DIQKLRLFAV LEQLSPVRAF QWDPQQPRLA ICTGGSRLYL WSPAGCMSVQ VPGE (201-413 aa encoded by B C002611)
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Studies of the related mouse protein suggest that the encoded protein may play a role in the process of ossification.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat domain 8; WD repeat-containing protein 8; WD repeat-containing protein antisense to TP73 gene; WD repeat-containing protein WRAP73
Gene Aliases: WDR8; WRAP73
UniProt ID: (Human) Q9P2S5
Entrez Gene ID: (Human) 49856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support