Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
WDR91 (WD repeat-containing 91) contains seven WD repeats and is a member of the WD family of proteins. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. WD proteins are involved in a variety of cellular processes. The function of WDR91 has not been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat-containing protein 91
Gene Aliases: 9530020G05Rik; AI987683; AU018665; HSPC049; RGD1306697; SORF-1; SORF1; WDR91
UniProt ID: (Human) A4D1P6, (Rat) B2RYI0, (Mouse) Q7TMQ7
Entrez Gene ID: (Human) 29062, (Rat) 312225, (Mouse) 101240
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support