Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSFLSRQQPP PPRRAGAACT LRQKLIFSPC SDCEEEEEEE EEEGSGHSTG EDSAFQEPDS PLPPARSPTE PGPERRRSPG PAPGSPGELE EDLLLPGACP
WEE1, which is classified as a Ser/Thr kinase by sequence analysis, has been shown to inactivate multiple CDKs by tyrosine phosphorylation. Wee1 protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686I18166; FLJ16446; WEE1 tyrosine kinase; Wee1-like protein kinase; Wee1A kinase
Gene Aliases: Wee1; Wee1A
UniProt ID: (Mouse) P47810
Entrez Gene ID: (Mouse) 22390
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support