Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TKSGNHFEEP KLKDILLQIS LGLNYIHNSS MVHLDIKPSN IFICHKMQSE SSGVIEEVEN EADWFLSANV MYKIGDLGHA TSINKPKVEE GD
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
Oocyte-specific protein tyrosine kinase that phosphorylates and inhibits CDK1/CDC2 and acts as a key regulator of meiosis during both prophase I and metaphase II (PubMed:29606300). Required to maintain meiotic arrest in oocytes during the germinal vesicle (GV) stage, a long period of quiescence at dictyate prophase I, by phosphorylating CDK1 at 'Tyr-15', leading to inhibit CDK1 activity and prevent meiotic reentry. Also required for metaphase II exit during egg activation by phosphorylating CDK1 at 'Tyr-15', to ensure exit from meiosis in oocytes and promote pronuclear formation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Wee1-like protein kinase 1B; Wee1-like protein kinase 2; Wee1B kinase
Gene Aliases: WEE1B; WEE2
UniProt ID: (Human) P0C1S8
Entrez Gene ID: (Human) 494551
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support