Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEFSIKQSPL SVQSVVKCIK MKQAPEILGS ANGKTPSCEV NRECSVFLSK AQLSSSLQEG VMQKFNGHDA LPFIPADKLK DLTSRVFNGE PGAHDAKLRF ESQEMKGIGT PPNTTPIKNG SPEIKLKITK TYMNGKPLFE SSICGDSAAD VSQSEENGQK PENKARRNRK RSIKYDSLLE QGLVEAALVS KISSPSDKKI PAKKESCPNT GRDKDHLLKY NVGDLVWSKV SGYPWWPCMV SADPLLHSYT KLKGQKKSAR QYHVQFFGDA PERAWIFEKS LVAFEGEGQF EKLCQESAKQ APTKAEKIKL LKPISGKLRA QWEMGIVQAE EAASMSVEER KAKFTFLYVG DQLHLNPQVA KEAGIAAESL GEMAESSGVS EEAAENPKSV REECIPMKRR RRAKLCSSAE TLESHPDIGK STPQKTAEAD PRRGVGSPPG RKKTTVSMPR SRKGDAASQF LVFCQKHRDE VVAEHPDASG EEIEELLRSQ WSLLSEKQRA RYNTKFALVA PVQAEEDSGN VNGKKRNHTK RIQDPTEDAE AEDTPRKRLR TDKHSLRKRD TITDKTARTS SYKAMEAASS LKSQAATKNL SDACKPLKKR NRASTAASSA LGFSKSSSPS ASLTENELLW EPTPVKLDLN PAALYCT
This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; MMSET; multiple myeloma SET domain containing protein type III; Multiple myeloma SET domain-containing protein; nuclear receptor binding SET domain protein 2; Nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; trithorax/ash1-related protein 5; Wolf-Hirschhorn syndrome candidate 1 protein
Gene Aliases: KIAA1090; MMSET; NSD2; REIIBP; TRX5; WHS; WHSC1
UniProt ID: (Human) O96028
Entrez Gene ID: (Human) 7468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support