Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
WIPI1 (WD repeat domain, phosphoinositide interacting-1), also known as WIPI1, ATG18 or WIPI49, is thought to play a role in autophagy and may regulate protein trafficking in certain recycling pathways. It contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. WIPI1 localizes to cytoplasmic vesicles, endosomes, clathrin-coated vesicles and the trans-Golgi network. It is ubiquitously expressed with highest expression in heart, testis, placenta, pancreas and skeletal muscle. WIPI1 is upregulated in a variety of tumors, suggesting a role in carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atg18 protein homolog; WD repeat domain phosphoinositide-interacting protein 1; WD40 repeat protein interacting with phosphoinositides of 49 kDa; WD40 repeat protein Interacting with phosphoInositides of 49kDa; WIPI 49 kDa; WIPI-1; WIPI-1 alpha
Gene Aliases: 4930533H01Rik; ATG18; ATG18A; AW411817; D11Ertd498e; WIPI1; WIPI49
UniProt ID: (Human) Q5MNZ9, (Mouse) Q8R3E3
Entrez Gene ID: (Human) 55062, (Mouse) 52639
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support