Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with mouse and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
WIZ, also known as ZNF803, was initially identified in a mouse cerebellum cDNA library screen and its message was found to be expressed in the granule cell layers of the cerebellum as well as in the dentate gyrus and olfactory bulb. Later analysis indicates however that WIZ is ubiquitously expressed. WIZ is a nuclear protein that co-localizes with G9a, a histone methyltransferase responsible for mono- and dimethylation of H3K9 at euchromatic regions. WIZ can also associate with CtBP family proteins, known to be transcriptional co-repressors, and has been suggested to link G9a/GLP complexes to the CtBP co-repressor machinery, possibly regulating complex stability and gene silencing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wiz; widely-interspaced zinc finger motifs; Widely-interspaced zinc finger-containing protein; WIZ zinc finger; Zinc finger protein 803
Gene Aliases: WIZ; ZNF803
UniProt ID: (Human) O95785
Entrez Gene ID: (Human) 58525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support