Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LKACGCDAS RRGDEEAFRR KLHRLQLDAL QRGKGLSHGV PEHPALPTAS PGLQDSWEWG GCSPDMGFGE RFSKDFL (154-229 aa encoded by BC052234 )
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is strongly expressed in the cell lines of promyelocytic leukemia and Burkitt's lymphoma. In addition, it and another family member, the WNT6 gene, are strongly coexpressed in colorectal cancer cell lines. The gene overexpression may play key roles in carcinogenesis through activation of the WNT-beta-catenin-TCF signaling pathway. This gene and the WNT6 gene are clustered in the chromosome 2q35 region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-10a; wingless related MMTV integration site 10a; wingless-type MMTV integration site family member 10A; wingless-type MMTV integration site family, member 10A
Gene Aliases: OODD; SSPS; STHAG4; WNT10A
UniProt ID: (Human) Q9GZT5, (Mouse) P70701
Entrez Gene ID: (Human) 80326, (Mouse) 22409
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support