Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNAPLGGIW LWLPLLLTWL TPEVNSSWWY MRATGGSSRV MCDNVPGLVS SQRQLCHRHP DVMRAISQGV AEWTAECQHQ FRQHRWNCNT LDRDHSLFGR VLLRSSRESA FVYAISSAGV VFAITRACSQ GEVKSCSCDP KKMGSAKDSK GIFDWGGCSD NIDYGIKFAR AFVDAKERKG KDARALMNLH NNRAGRKAVK RFLKQECKCH GVSGSCTLRT CWLAMADFRK TGDYLWRKYN GAIQVVMNQD GTGFTVANER FKKPTKNDLV YFENSPDYCI RDREAGSLGT AGRVCNLTSR GMDSCEVMCC GRGYDTSHVT RMTKCGCKFH WCCAVRCQDC LEALDVHTCK APKNADWTTA T (1-360 aa encoded by BC029854)
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Int-1-like protein 1; INT-1-related protein; INT1L 1; IRP; IRP protein; Protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; WNT 2
Gene Aliases: 2610510E18Rik; INT1L1; IRP; Mirp; Wnt; Wnt-2; WNT2; Wnt2a
UniProt ID: (Human) P09544, (Mouse) P21552
Entrez Gene ID: (Human) 7472, (Mouse) 22413, (Rat) 114487
Molecular Function: intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support