Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRPPPALALA GLCLLALPAA AASYFGLTGR EVLTPFPGLG TAAAPAQGGA HLKQCDLLKL SRRQKQLCRR EPGLAETLRD AAHLGLLECQ FQFRHERWNC SLEGRTGLLK RGFKETAFLY AVSSAALTHT LARACSAGRM ERCTCDDSPG LESRQAWQWG VCGDNLKYST KFLSNFLGSK RGNKDLRARA DAHNTHVGIK AVKSGLRTTC KCHGVSGSCA VRTCWKQLSP FRETGQVLKL RYDSAVKVSS ATNEALGRLE LWAPARQGSL TKGLAPRSGD LVYMEDSPSF CRPSKYSPGT AGWSAVARSQ LITTSTSRIQ AILPSQLPE
WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15
Gene Aliases: UNQ6973/PRO21956; WNT14B; WNT15; WNT9B
UniProt ID: (Human) O14905
Entrez Gene ID: (Human) 7484
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support