Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-6; wingless-related MMTV integration site 6; wingless-type MMTV integration site family member 6; wingless-type MMTV integration site family, member 6
Gene Aliases: AA409270; Wnt-6; WNT6
UniProt ID: (Human) Q9Y6F9, (Mouse) P22727
Entrez Gene ID: (Human) 7475, (Rat) 316526, (Mouse) 22420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support