Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGDFSVCRN CKRHVVSANF TLHEAYCLRF LVLCPECEEP VPKETMEEHC KLEHQQVGCT MCQQSMQKSS LEFHKANECQ ERPVECKFCK LDMQLSKLEL HESYCGSRTE LCQGCGQFIM HRMLAQHRDV CRSEQAQLGK GERISAPERE IYCHYCNQMI PENKYFHHMG KCCPDSEFKK HFPVGNPEIL PSSLPSQAAE NQTSTMEKDV RPKTRSINRF PLHSESSSKK APRSKNKTLD PLLMSEPKPR TSSPRGDKAA YDILRRCSQC GILLPLPILN QHQEKCRWLA SSKGKQVRNF S
The inhibitors of apoptosis (IAP) proteins prevent cell death both in vitro and in vivo by virtue of inhibition of distinct caspases. The X-linked Inhibitor of Apoptosis, XIAP, is a key member of the IAP family of proteins. The caspase inhibiting activity of XIAP is negatively regulated by at least two XIAP-interacting proteins, XIAP-associated factor 1 (XAF1) and Smac/DIABLO. XAF1 binds to XIAP and antagonizes caspase inhibitory activities of XIAP. Expression of XAF1 triggers a redistribution of XIAP from the cytosol to the nucleus. XAF1 is ubiquitously expressed in normal tissues, but is present at low or undetectable levels in many different cancer cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BIRC4 binding protein; BIRC4-binding protein; F-box protein 39; novel TRAF-type zinc finger protein; XIAP-associated factor 1
Gene Aliases: BIRC4BP; Fbox39; HSXIAPAF1; XAF1; XIAPAF1
UniProt ID: (Human) Q6GPH4, (Mouse) Q5NBU8
Entrez Gene ID: (Human) 54739, (Mouse) 327959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support