Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total XCL1/2.
Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATAC; C motif chemokine 1; C motif chemokine 2; chemokine (C motif) ligand 1; chemokine (C motif) ligand 2; Cytokine SCM-1; Cytokine SCM-1 beta; Lymphotactin; Lymphotaxin; SCM-1-alpha; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small inducible cytokine subfamily C, member 2; Small-inducible cytokine C1; XC chemokine ligand 1; XC chemokine ligand 2
Gene Aliases: AI661682; ATAC; LPTN; LTN; SCM-1; SCM-1a; SCM-1b; SCM1; SCM1A; SCM1B; SCYC1; SCYC2; XCL1; XCL2
UniProt ID: (Human) P47992, (Human) Q9UBD3, (Mouse) P47993
Entrez Gene ID: (Human) 6375, (Human) 6846, (Mouse) 16963
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support