Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: PCSTSYCCLA PRTFGVSPLR RWIQRPQNCN TGSSGCCENS GSSGCCGSGG CGCSCGCGSS G
Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 71%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KPRP N-terminal and LCE C-terminal-like protein; Protein KPLCE; skin-specific protein (xp32); Skin-specific protein 32
Gene Aliases: C1orf68; KPLCE; LEP7; XP32
UniProt ID: (Human) Q5T750
Entrez Gene ID: (Human) 100129271
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support