Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAADGALPE AAALEQPAEL PASVRASIER KRQRALMLRQ ARLAARPYSA TAAAATGGMA NVKAAPKIID TGGGFILEEE EEEEQKIGKV VHQPGPVMEF DYVICEECGK EFMDSYLMNH FDLPTCDNCR DADDKHKLIT KTEAKQEYLL KDCDLEKREP PLKFIVKKNP HHSQWGDMKL YLKLQIVKRS LEVWGSQEAL EEAKEVRQEN REKMKQKKFD KKVKELRRAV RSSVWKRETI VHQHEYGPEE NLEDDMYRKT CTMCGHELTY EKM
The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA repair protein complementing XP-A cells; excision repair-controlling; mutant xeroderma pigmentosum complementation group A; Xeroderma pigmentosum group A-complementing protein; xeroderma pigmentosum, complementation group A
Gene Aliases: XP1; XPA; XPAC
UniProt ID: (Human) P23025
Entrez Gene ID: (Human) 7507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support