Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F
Gene Aliases: AI606920; ERCC11; ERCC4; FANCQ; RAD1; RGD1560340; XFEPS; XPF
UniProt ID: (Human) Q92889, (Mouse) Q9QZD4
Entrez Gene ID: (Human) 2072, (Dog) 479842, (Mouse) 50505, (Rat) 304719
Molecular Function:
DNA metabolism protein
endodeoxyribonuclease
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support