Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EFPGHGLEP LLKFSEWWHE AQKNPKIRPN PHDTKVKKKL RTLQLTPGFP NPAVAEAYLK PVVDDSKGSF LWGKPDLDKI REFCQRYFGW NRTKTDESLF PVLKQLDAQQ TQLRIDSFFR LAQQEKEDAK RIKSQRLNRA VTCMLRKEKE AAASEIEAVS VAMEKEFELL DKAKRKTQKR GITNTLEESS SLKRKRLSDS KRKNTCGGFL GETCLSESSD GSSSEDAESS SLMNVQRRTA AKEPKTSASD SQNSVKEAPV KNGGATTSSS SDSDDDGGKE KMVLVTARSV FGKKRRKLRR ARGRKRKT (880-1186 aa encoded by BC031522)
Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COFS 3; DNA excision repair protein ERCC-5; DNA repair protein complementing XP-G cells; ERCC 5; ERCM 2; excision repair cross-complementation group 5; excision repair cross-complementing rodent repair deficiency, complementation group 5; OTTHUMP00000064902; RP11-484I6.5; Xe; xeroderma pigmentosum; Xeroderma pigmentosum group G-complementing protein; xeroderma pigmentosum, complementation group G; XPG-complementing protein
Gene Aliases: COFS3; ERCC5; ERCC5-201; ERCM2; UVDR; XPG; XPGC
UniProt ID: (Human) P28715
Entrez Gene ID: (Human) 2073
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support