Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: PVPERRIPNR YLGQPSPFTH PHLLRPGEVT PGLSQVEYAL RRHKLMSLIQ
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminopeptidase P3; APP3; Intermediate Cleaving Peptidase 55; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3, mitochondrial; Xaa-Pro aminopeptidase 3
Gene Aliases: APP3; ICP55; NPHPL1; XPNPEP3
UniProt ID: (Human) Q9NQH7
Entrez Gene ID: (Human) 63929
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support