Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminopeptidase P3; APP3; Intermediate Cleaving Peptidase 55; probable Xaa-Pro aminopeptidase 3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3, mitochondrial; Xaa-Pro aminopeptidase 3
Gene Aliases: APP3; E430012M05Rik; ICP55; NPHPL1; XPNPEP3
UniProt ID: (Human) Q9NQH7, (Rat) B5DEQ3, (Mouse) B7ZMP1
Entrez Gene ID: (Dog) 481237, (Human) 63929, (Rat) 685823, (Mouse) 321003
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support