Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDEQALLGLN PNADSDFRQR ALAYFEQLKI SPDAWQVCAE ALAQRTYSDD HVKFFCFQVL EHQVKYKYSE LTTVQQQLIR ETLISWLQAQ MLNPQPEKTF IRNKAAQVFA LLFVTEYLTK WPKFFFDILS VVDLNPRGVD LYLRILMAID SELVDRDVVH TSEEARRNTL IKDTMREQCI PNLVESWYQI LQNYQFTNSE VTCQCLEVVG AYVSWIDLSL IANDRFINML LGHMSIEVLR EEACDCLFEV VNKGMDPVDK MKLVESLCQV LQSAGFFSID QEEDVDFLAR FSKLVNGMGQ SLIVSWSKLI KNGDIKNAQE ALQAIETKVA LMLQLLIHED DDISSNIIGF CYDYLHILKQ LTVLSDQQKA NVEAIMLAVM KKLTYDEEYN FENEGEDEAM FVEYRKQLKL LLDRLAQVSP ELLLASVRRV FSSTLQNWQT TRFMEVEVAI RLLYMLAEAL PVSHGAHFSG DVSKASALQD MMRTLVTSGV SSYQHTSVTL EFFETVVRYE KFFTVEPQHI PCVLMAFLDH RGLRHSSAKV RSRTAYLFSR FVKSLNKQMN PFIEDILNRI QDLLELSPPE NGHQSLLSSD DQLFIYETAG VLIVNSEYPA ERKQALMRNL LTPLMEKFKI LLEKLMLAQD EERQASLADC LNHAVGFASR TSKAFSNKQT VKQCGCSEVY LDCLQTFLPA LSCPLQKDIL RSGVRTFLHR MIICLEEEVL PFIPSASEHM LKDCEAKDLQ EFIPLINQIT AKFKIQVSPF LQQMFMPLLH AIFEVLLRPA EENDQSAALE KQMLRRSYFA FLQTVTGSGM SEVIANQGAE NVERVLVTVI QGAVEYPDPI AQKTCFIILS KLVELWGGKD GPVGFADFVY KHIVPACFLA PLKQTFDLAD AQTVLALSEC AVTLKTIHLK RGPECVQYLQ QEYLPSLQVA PEIIQEFCQA LQQPDAKVFK NYLKVFFQRA KP
XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exportin(tRNA); exportin, tRNA (nuclear export receptor for tRNAs); Exportin-T; tRNA exportin
Gene Aliases: XPO3; XPOT
UniProt ID: (Human) O43592
Entrez Gene ID: (Human) 11260
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support