Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSGWESYYKT EGDEEAEEEQ EENLEASGDY KYSGRDSLIF LVDASKAMFE SQSEDELTPF DMSIQCIQSV YISKIISSDR DLLAVVFYGT EKDKNSVNFK NIYVLQELDN PGAKRILELD QFKGQQGQKR FQDMMGHGSD YSLSEVLWVC ANLFSDVQFK MSHKRIMLFT NEDNPHGNDS AKASRARTKA GDLRDTGIFL DLMHLKKPGG FDISLFYRDT ISIAEDEDLR VHFEESSKLE DLLRKVRAKE TRKRALSRLK LKLNKDIVIS VGIYNLVQKA LKPPPIKLYR ETNEPVKTKT RTFNTSTGGL LLPSDTKRSQ IYGSRQIILE KEETEELKRF DDPGLMLMGF KPLVLLKKHH YLRPSLFVYP EESLVIGSST LFSALLIKCL EKEVAALCRY TPRRNIPPYF VALVPQEEEL DDQKIQVTPP GFQLVFLPFA DDKRKMPFTE KIMATPEQVG KMKAIVEKLR FTYRSDSFEN PVLQQHFRNL EALALDLMEP EQAVDLTLPK VEAMNKRLGS LVDEFKELVY PPDYNPEGKV TKRKHDNEGS GSKRPKVEYS EEELKTHISK GTLGKFTVPM LKEACRAYGL KSGLKKQELL EALTKHFQD
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 70 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CT; CTA-216E10.7; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen, 70kDa; Ku70; Lupus Ku autoantigen protein p70; OTTHUMP00000028581; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 6
Gene Aliases: CTC75; CTCBF; G22P1; KU70; ML8; TLAA; XRCC6
UniProt ID: (Human) P12956
Entrez Gene ID: (Human) 2547
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support