Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31955 has been tested in indirect ELISA.
XRN2 gene encodes a 5'-3' exonuclease that promotes transcription termination at cotranscriptional cleavage sites. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-3' exoribonuclease 2; DHM1-like protein; DHP protein; XRN2b
Gene Aliases: XRN2
UniProt ID: (Human) Q9H0D6
Entrez Gene ID: (Human) 22803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support