Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Involved in the maintenance of the Golgi structure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein YIPF4; putative protein product of Nbla11189; Yip1 domain family, member 4; YIP1 family member 4
Gene Aliases: FinGER4; Nbla11189; YIPF4
UniProt ID: (Human) Q9BSR8
Entrez Gene ID: (Human) 84272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support