Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis and Bovine
This gene product is one of the SNARE recognition molecules implicated in vesicular transport between secretory compartments. It is a membrane associated, isoprenylated protein that functions at the endoplasmic reticulum-Golgi transport step. This protein is highly conserved from yeast to human and can functionally complement the loss of the yeast homolog in the yeast secretory pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: prenylated SNARE protein (Ykt6); R-SNARE; SNARE protein Ykt6; Synaptobrevin homolog YKT6; YKT6 v-SNARE protein; YKT6, S. cerevisiae, homolog of
Gene Aliases: 0610042I15Rik; 1810013M05Rik; AW105923; YKT6
UniProt ID: (Human) O15498, (Mouse) Q9CQW1
Entrez Gene ID: (Human) 10652, (Mouse) 56418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support