Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LSSRTRVRE LQGQIAAITG IAPGGQRILV GYPPECLDLS NGDTILEDLP IQSGDMLIIE EDQTRPRSSP AFTKRGASSY VRETLPVLTR TVVPADNSCL FTSVYYVVEG GVLNPACAPE MRRLIAQIVA SDPDFYSEAI LGKTNQEYCD WIKRDDTWGG AIEISILSKF YQCEICVVDT QTVRIDRFGE DAGYTKRVLL IYDGIHYDPL QRNFPDPDTP PLTIFSSNDD IVLVQALELA DEARRRRQFT DVNRFTLRCM VCQKGLTGQA EAREHAKETG HTNFGEV (63-348 aa encoded by BC137167)
Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DUBA-8; HIN-7; HIV-1-induced protease 7; hsHIN7; OTU domain containing 2; OTU domain-containing protein 2; Ubiquitin thioesterase OTU1; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast)
Gene Aliases: DUBA8; HIN7; OTUD2; PRO0907; YOD1
UniProt ID: (Human) Q5VVQ6
Entrez Gene ID: (Human) 55432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support