Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521-B; YTH domain-containing protein 1
Gene Aliases: A730098D12Rik; C80342; KIAA1966; mKIAA1966; YT521; YT521-B; YTHDC1
UniProt ID: (Human) Q96MU7, (Mouse) E9Q5K9, (Rat) Q9QY02
Entrez Gene ID: (Human) 91746, (Mouse) 231386, (Rat) 170956
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support