Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MASGDTLYIA TDGSEMPAEI VELHEIEVET IPVETIETTV VGEEEEEDDD DEDGGGGDHG GGGGHGHAGH HHHHHHHHHH PPMIALQPLV TDDPTQVHHH QEVILVQTRE EVVGGDDSDG LRAEDGFEDQ ILIPVPAPAG GDDDYIEQTL VTVAAAGKSG GGGSSSSGGG RVKKGGGKKS GKKSYLSGGA GAAGGGGADP GNKKWEQKQV QIKTLEGEFS VTMWSSDEKK DIDHETVVEE QIIGENSPPD YSEYMTGKKL PPGGIPGIDL SDPKQLAEFA RMKPRKIKED DAPRTIACPH KGCTKMFRDN SAMRKHLHTH GPRVHVCAEC GKAFVESSKL KRHQLVHTGE KPFQCTFEGC GKRFSLDFNL RTHVRIHTGD RPYVCPFDGC NKKFAQSTNL KSHILTHAKA KNNQ
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delta transcription factor; INO80 complex subunit S; NF-E1; Transcriptional repressor protein YY1; UCR-motif DNA-binding protein; UCRBP transcription factor; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YY-1
Gene Aliases: AW488674; DELTA; INO80S; NF-E1; UCRBP; YIN-YANG-1; YY1
UniProt ID: (Human) P25490, (Mouse) Q00899
Entrez Gene ID: (Human) 7528, (Mouse) 22632
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support