Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
ZACN is a zinc-activated ligand-gated ion channel that defines a new subgroup of the cysteine-loop superfamily of ligand-gated ion channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ligand-gated cation channel ZACN; ligand-gated ion channel subunit; Ligand-gated ion channel zinc-activated 1; ligand-gated ion channel, zinc activated 1; Ligand-gated ion-channel receptor L2; zinc activated ligand-gated ion channel 1; Zinc-activated channel
Gene Aliases: L2; LGICZ; LGICZ1; ZAC; ZAC1; ZACN
UniProt ID: (Human) Q401N2
Entrez Gene ID: (Human) 353174
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support