Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Plays a role in the reduction of telomerase activity during differentiation of embryonic stem cells by binding to the core promoter of TERT and controlling its down-regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear protein ZAP3; protein phosphatase 1, regulatory subunit 169; YLP motif-containing protein 1; ZAP113
Gene Aliases: C14orf170; PPP1R169; YLPM1; ZAP113; ZAP3
UniProt ID: (Human) P49750
Entrez Gene ID: (Human) 56252
Molecular Function:
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support