Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This maternal effect gene is oocyte-specific and encodes a protein that is thought to function in the initiation of embryogenesis. A similar protein in mouse is required for female fertility. [provided by RefSeq, Jul 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oocyte-specific maternal effect factor; zinc finger, 3CxxC-type 6; Zygote arrest protein 1
Gene Aliases: Z3CXXC6; ZAR1
UniProt ID: (Human) Q86SH2, (Mouse) Q80SU3, (Rat) Q7TSX9
Entrez Gene ID: (Human) 326340, (Mouse) 317755, (Rat) 353228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support