Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMRREDEEEE GTMMKAKGDL EMKEEEEISE TGELVGPFVS AMPTPMPHNK GTRFSEAWEY FHLAPARAGH HPNQYATCRL CGRQVSRGPG VNVGTTALWK HLKSMHREEL EKSGHGQAGQ RQDPRPHGPQ LPTGIEGNWG RLLEQVGTMA LWASQREKEV LRRERAVEWR ERAVEKRERA LEEVERAILE MKWKVRAEKE ACQREKELPA AVHPFHFV
The ZBED2 protein possesses a BED-type zinc finger, a 50 to 60 amino acid domain that contains a characteristic motif with two highly conserved aromatic positions, as well as a shared pattern of cysteines and histidines that is predicted to form the zinc finger. ZBED2 was identified through a screening of 37 human gastric cancer cDNA libraries. While the function of this protein unknown at this time, the presence of the zinc finger suggests that it may have DNA or RNA binding properties.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger BED domain-containing protein 2; zinc finger, BED domain containing 2; zinc finger, BED-type containing 2
Gene Aliases: ZBED2
UniProt ID: (Human) Q9BTP6
Entrez Gene ID: (Human) 79413
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support