Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is unusual in that its coding sequence is mostly derived from Charlie-like DNA transposon; however, it does not appear to be an active DNA transposon as it is not flanked by terminal inverted repeats. The encoded protein is conserved among the mammalian Laurasiatheria branch. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transposon-derived Buster1 transposase-like protein; Zinc finger BED domain-containing protein 5; zinc finger, BED-type containing 5
Gene Aliases: Buster1; ZBED5
UniProt ID: (Human) Q49AG3
Entrez Gene ID: (Human) 58486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support