Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAKPSHSSY VLQQLNNQRE WGFLCDCCIA IDDIYFQAHK AVLAACSSYF RMFFMNHQHS TAQLNLSNMK ISAECFDLIL QFMYLGKIMT APSSFEQFKV AMNYLQLYNV PDCLEDIQDA DCSSSKCSSS ASSKQNSKMI FGVRMYEDTV ARNGNEANRW CAEPSSTVNT PHNREADEES LQLGNFPEPL FDVCKKSSVS KLSTPKERVS RRFGRSFTCD SCGFGFSCEK LLDEHVLTCT NRHLYQNTRS YHRIVDIRDG KDSNIKAEFG EKDSSKTFSA QTDKYRGDTS QAADDSASTT GSRKSSTVES EIASEEKSRA AERKRIIIKM EPEDTPTDEL KDFNIIKVTD KDCNESTD (1-357 aa encoded by BC050719)
ZBTB1 acts as a transcriptional repressor. Represses cAMP-responsive element (CRE)-mediated transcriptional activation. In addition, has a role in translesion DNA synthesis. Requires for UV-inducible RAD18 loading, PCNA monoubiquitination, POLH recruitment to replication factories and efficient translesion DNA synthesis. Plays a key role in the transcriptional regulation of T lymphocyte development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 1
Gene Aliases: KIAA0997; ZBTB1; ZNF909
UniProt ID: (Human) Q9Y2K1
Entrez Gene ID: (Human) 22890
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support