Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGMGNDISAE DICAEDIPKH RQKVDQPLKD QENLVASTAK TDFGPDDDTY RSRLRQRSVN EGAYIRLHKG MEKKLQKRKA VPKSAVQQVA QKLVQRGKKM KQPKRDAKEN TEEASH
Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 11
Gene Aliases: ZBTB11; ZNF-U69274; ZNF913
UniProt ID: (Human) O95625
Entrez Gene ID: (Human) 27107
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support