Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GLGIGGSVGG HLGELAQSSV PPSTVAPPQG VVKACY
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 89%.
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLA-B-associated transcript 9; Protein G10; Zinc finger and BTB domain-containing protein 12
Gene Aliases: Bat9; C6orf46; D6S59E; G10; NG35; ZBTB12
UniProt ID: (Human) Q9Y330
Entrez Gene ID: (Human) 221527
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support