Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein BING1; Zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; Zinc finger protein 297
Gene Aliases: 1110008J20Rik; AI265210; AI415166; BING1; fru; fruitless; ZBTB22; ZBTB22A; Zfp297; ZNF297; ZNF297A
UniProt ID: (Human) O15209, (Mouse) Q9Z0G7
Entrez Gene ID: (Human) 9278, (Mouse) 81630, (Rat) 309630
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support