Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLPPIRLPS PYGSDRLVQL AARLRPALCD TLITVGSQEF PAHSLVLAGV SQQLGRRGQW ALGEGISPST FAQLLNFVYG ESVELQPGEL RPLQEAARAL GVQSLEEACW RARGDRAKKP DPGLKKHQEE PEKPSRNPER ELGDPGEKQK PKQVSRTGGR EQEMLHKHSP PRGRPEMAGA TQEAQQEQTR SKEKRLQAPV GQRGADGKHG VLTWLRENPG GSEESLRKLP GPLPPAGSLQ TSVTPRPSWA EAPWLVGGQP ALWSILLMPP RYGIPFYHST PTTGAWQEVW REQRRTCNLC GS
DNA-binding protein that binds to the to a 5'-TGTACAGTGT-3' core sequence. May function as a transcriptional transactivator and transcriptional repressor. Probably exerts its repressor effect by preventing GATA3 from binding to DNA. May play a role in regulating the differentiation and activation of helper T-cells. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FANCC-interacting protein; Fanconi anemia zinc finger protein; OTTHUMP00000045790; repressor of GATA; Testis zinc finger protein; Zinc finger and BTB domain-containing protein 32; Zinc finger protein 538
Gene Aliases: FAXF; FAZF; Rog; TZFP; ZBTB32; ZNF538
UniProt ID: (Human) Q9Y2Y4
Entrez Gene ID: (Human) 27033
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support