Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEKGGNIQLE IPDFSNSVLS HLNQLRMQGR LCDIVVNVQG QAFRAHKVVL AASSPYFRDH MSLNEMSTVS ISVIKNPTVF EQLLSFCYTG RICLQLADII SYLTAASFLQ MQHIIDKCTQ ILEGIHFKIN VAEVEAELSQ TRTKHQERPP ESHRVTPNLN RSLSPRHNTP KGNRRGQVSA VLDIRELSPP EESTSPQIIE PSSDVESREP ILRINRAGQW YVETGVADRG GRSDDEVRVL GAVHIKTENL EEWLGPENQP SGEDGSSAEE VTAMVIDTTG HGSVGQENYT LGSSGAKVAR PTSSEVDRFS PSGSVVPLTE RHRARSESPG RMDEPKQPSS QVWSCGFRTA LVVGGIATVY E
ZBTB37 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 37
Gene Aliases: D430004I08Rik; ZBTB37; ZNF908
UniProt ID: (Human) Q5TC79
Entrez Gene ID: (Human) 84614
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support