Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22332; FLJ31131; FLJ35036; protein phosphatase 1, regulatory subunit 171; ZBTB38; Zinc finger and BTB domain-containing protein 38
Gene Aliases: CIBZ; PPP1R171; ZBTB38; ZNF921
UniProt ID: (Human) Q8NAP3
Entrez Gene ID: (Human) 253461
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support