Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGMRIKLQST NHPNNLLKEL NKCRLSETMC DVTIVVGSRS FPAHKAVLAC AAGYFQNLFL NTGLDAARTY VVDFITPANF EKVLSFVYTS ELFTDLINVG VIYEVAERLG MEDLLQACHS TFPDLESTAR AKPLTSTSES HSGTLSCPSA EPAHPLGELR GGGDYLGADR NYVLPSDAGG SYKEEEKNVA SDANHSLHLP QPPPPPPKTE DHDTPAPFTS IPSMMTQPLL GTVSTGIQTS TSSCQPYKVQ SNGDFSKNSF LTPDNAVDIT TGTNSCLSNS EHSKDPGFGQ MDELQLEDLG DDDLQFEDPA EDIGTTEEVI ELSDDSEDEL AFGENDNREN KAMPCQVCKK VLEPNIQLIR QHARDHVDLL TGNCKVCETH FQDRNSRVTH VLSHIGIFLF SCDMCETKFF TQWQLTLHRR DGIFENNIIV HPNDPLPGKL GLFSGAASPE LKCAACGKVL AKDFHVVRGH ILDHLNLKGQ ACSVCDQRHL NLCSLMWHTL SHLGISVFSC SVCANSFVDW HLLEKHMAVH QSLEDALFHC RLCSQSFKSE AAYRYHVSQH KCNSGLDARP GFGLQHPALQ KRKLPAEEFL GEELALQGQP GNSKYSCKVC GKRFAHTSEF NYHRRIHTGE KPYQCKVCHK FFRGRSTIKC HLKTHSGALM YRCTVCGHYS STLNLMSKHV GVHKGSLPPD FTIEQTFMYI IHSKEADKNP DS
ZBTB39 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 39
Gene Aliases: KIAA0352; ZBTB39; ZNF922
UniProt ID: (Human) O15060
Entrez Gene ID: (Human) 9880
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support