Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FRBZ1 protein (FRBZ1); RP11-469L3.1; zinc finger and BTB domain containing 41 homolog; Zinc finger and BTB domain-containing protein 41
Gene Aliases: 8430415N23Rik; 9430031N01; 9830132G07Rik; AI316857; FRBZ1; RGD1560834; ZBTB41; ZNF924
UniProt ID: (Human) Q5SVQ8, (Mouse) Q811F1
Entrez Gene ID: (Human) 360023, (Rat) 289052, (Mouse) 226470
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support