Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METPTPLPPV PASPTCNPAP RTIQIEFPQH SSSLLESLNR HRLEGKFCDV SLLVQGRELR AHKAVLAAAS PYFHDKLLLG DAPRLTLPSV IEADAFEGLL QLIYSGRLRL PLDALPAHLL VASGLQMWQV VDQCSEILRE LETSGGGISA RGGNSYHALL STTSSTGGWC IRSSPFQTPV QSSASTESPA STESPVGGEG SELGEVLQIQ VEEEEEEEED DDDEDQGSAT LSQTPQPQRV SGVFPRPHGP HPLPMTATPR KLPEGESAPL ELPAPPALPP KIFYIKQEPF EPKEEISGSG TQPGGAKEET KVFSGGDTEG NGELGFLLPS GPGPTSGGGG PSWKPVDLHG NEILSGGGGP GGAGQAVHGP VKLGGTPPAD GKRFGCLCGK RFAVKPKRDR HIMLTFSLRP FGCGICNKRF KLKHHLTEHM KTHAGALHAC PHCGRRFRVH ACFLRHRDLC KGQGWATAHW TYK
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB9, also known as ZNF919, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB9, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB9 may act as a transcriptional repressor or be involved in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DASS-97D12.7; Zinc finger and BTB domain-containing protein 9
Gene Aliases: ZBTB9; ZNF919
UniProt ID: (Human) Q96C00
Entrez Gene ID: (Human) 221504
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support