Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPNQGEDCYF FFYSTCTKGD SCPFRHCEAA IGNETVCTLW QEGRCFRQVC RFRHMEIDKK RSEIPCYWEN QPTGCQKLNC AFHHNRGRYV DGLFLPPSKT VLPTVPESPE EEVKASQLSV QQNKLSVQSN PSPQLRSVMK VESSENVPSP THPPVVINAA DDDEDDDDQF SEEGDETKTP TLQPTPEVHN GLRVTSVRKP AVNIKQGECL NFGIKTLEEI KSKKMKEKSK KQGEGSSGVS SLLLHPEPVP GPEKENVRTV VRTVTLSTKQ GEEPLVRLSL TERLGKRKFS AGGDSDPPLK RSLAQRLGKK VEAPETNIDK TPKKAQVSKS LKERLGMSAD PDNEDATDKV NKVGEIHVKT LEEILLERAS QKRGELQTKL KTEGPSKTDD STSGARSSST IRIKTFSEVL AEKKHRQQEA ERQKSKKDTT CIKLKIDSEI KKTVVLPPIV ASRGQSEEPA GKTKSMQEVH IKTLEEIKLE KALRVQQSSE SSTSSPSQHE ATPGARRLLR ITKRTGMKEE KNLQEGNEVD SQSSIRTEAK EASGETTGVD ITKIQVKRCE TMREKHMQKQ QEREKSVLTP LRGDVASCNT QVAEKPVLTA VPGITRHLTK RLPTKSSQKV EVETSGIGDS LLNVKCAAQT LEKRGKAKPK VNVKPSVVKV VSSPKLAPKR KAVEMHAAVI AAVKPLSSSS VLQEPPAKKA AVAVVPLVSE DKSVTVPEAE NPRDSLVLPP TQSSSDSSPP EVSGPSSSQM SMKTRRLSSA STGKPPLSVE DDFEKLIWEI SGGKLEAEID LDPGKDEDDL LLELSEMIDS
Involved in nuclear mRNA export; probably mediated by association with the TREX complex. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH-type domain containing 11A
Gene Aliases: KIAA0663; RGD1308290; ZC3H11A; ZC3HDC11A
UniProt ID: (Human) O75152
Entrez Gene ID: (Human) 9877, (Rat) 360845
Molecular Function: RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support