Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat brain tissue lysate.
PA5-20928 can be used with blocking peptide PEP-1042.
ZC3H12C, also known as MCPIP3, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. Similar to ZC3H12A, ZC3H12C expression in macrophages is highly increased after treatment with lipopolysaccharide (LPS), suggesting it also may play a role in host immunity and inflammatory response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCP induced protein 3; MCP-induced protein 3; Probable ribonuclease ZC3H12C; Zinc finger CCCH domain-containing protein 12C; zinc finger CCCH-type containing 12C
Gene Aliases: A230108E06; C230027N18Rik; KIAA1726; MCPIP3; mKIAA1726; RGD1565078; ZC3H12C
UniProt ID: (Human) Q9C0D7, (Mouse) Q5DTV4
Entrez Gene ID: (Human) 85463, (Mouse) 244871, (Rat) 315658
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support