Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
ZC3H18 contains 1 C3H1-type zinc finger.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: conserved nuclear protein NHN1; Nuclear protein NHN1; Zinc finger CCCH domain-containing protein 18
Gene Aliases: NHN1; ZC3H18
UniProt ID: (Human) Q86VM9
Entrez Gene ID: (Human) 124245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support